Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromotri(4-biphenylyl)amine 98.0+%, TCI America™
SDP

Catalog No. B50201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50201G 1 g
B50205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50201G Supplier TCI America Supplier No. B50201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 728039-63-2
Molecular Formula C36H26BrN
Molecular Weight (g/mol) 552.52
MDL Number MFCD22571699
InChI Key VBQIVFTUXFKGKW-UHFFFAOYSA-N
PubChem CID 58672377
IUPAC Name N,N-bis({[1,1'-biphenyl]-4-yl})-4'-bromo-[1,1'-biphenyl]-4-amine
SMILES BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 1 g
Synonym N,N-Di(4-biphenylyl)-N-(4′C-bromo-4-biphenylyl)amine
Formula Weight 552.52
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromotri(4-biphenylyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.