Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxybenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. B104225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B104225G 25 g
B10425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B104225G Supplier TCI America Supplier No. B104225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6214-45-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
MDL Number MFCD00004657
InChI Key MGKNBDBZIKPUFM-UHFFFAOYSA-N
Synonym 4-butoxybenzyl alcohol, 4-butoxyphenyl methanol, 4-n-butoxybenzyl alcohol, p-butoxybenzyl alcohol, benzenemethanol, 4-butoxy, 4-butoxybenzylalcohol, benzenemethanol,4-butoxy, 4-butoxy-phenyl-methanol, 4-butoxyphenyl methanol #, acmc-1b65d
PubChem CID 80346
IUPAC Name (4-butoxyphenyl)methanol
SMILES CCCCOC1=CC=C(CO)C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 180.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Butoxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.