Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. B38885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38885G 5 g
B388825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38885G Supplier TCI America Supplier No. B38885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37909-95-8
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00100665
InChI Key JZDZRFNMDCBTNS-UHFFFAOYSA-N
Synonym 4-butyl-1,1'-biphenyl, 4-butylbiphenyl, 4-n-butylbiphenyl, 1,1'-biphenyl, 4-butyl, 4-butyl biphenyl, unii-1ty72t98fq, 1-p-biphenylylbutane, biphenyl, 4-butyl, 1-butyl-4-phenyl-benzene, 4-butyldiphenyl
PubChem CID 2775237
IUPAC Name 1-butyl-4-phenylbenzene
SMILES CCCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 129°C
Quantity 5 g
Formula Weight 210.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.