Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Carboxybenzo-15-crown 5-Ether 98.0+%, TCI America™
SDP

Catalog No. C17131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17131G Supplier TCI America Supplier No. C17131G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56683-55-7
Molecular Formula C15H20O7
Molecular Weight (g/mol) 312.318
MDL Number MFCD00143276
InChI Key FBNLTQGIRRAGRY-UHFFFAOYSA-N
Synonym (Benzo-15-crown 5-Ether)-4′C-carboxylic Acid, 2,3-(4-Carboxybenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 2724800
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carboxylic acid
SMILES C1COCCOC2=C(C=C(C=C2)C(=O)O)OCCOCCO1

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 312.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Carboxybenzo-15-crown 5-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.