Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1,2-phenylenediamine 97.0+%, TCI America™
SDP

Catalog No. C025525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025525G 25 g
C02555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025525G Supplier TCI America Supplier No. C025525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N

Specifications

Melting Point 75°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 142.59
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 4-Chloro-1,2-phenylenediamine
EINECSNumber (3)-0267
RTECSNumber SS8850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.