Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1,8-naphthalic anhydride, 94%

Catalog No. AAL0550822
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0550822 100 g
AAL0550814 25 g
AAL0550836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0550822 Supplier Thermo Scientific Chemicals Supplier No. L0550822
Add to Cart

CAS: 4053-08-1 | C12H5ClO3 | 232.62 g/mol

4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4053-08-1
Molecular Formula C12H5ClO3
Molecular Weight (g/mol) 232.62
MDL Number MFCD00006928
InChI Key UJEUBSWHCGDJQU-UHFFFAOYSA-N

Specifications

Melting Point 208°C to 214°C
Quantity 100 g
Beilstein 178072
Sensitivity Moisture sensitive
Synonym 4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione
Solubility Information Hydrolyzes in water.
SMILES C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl
PubChem CID 77672
Formula Weight 232.63
Percent Purity 94%
Chemical Name or Material 4-Chloro-1,8-naphthalic anhydride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-760-7
RTECSNumber QL6127295
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.