Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1H-pyrrolo[2,3-b]pyridine 97.0+%, TCI America™
SDP

Catalog No. C24701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24701G 1 g
C2470200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24701G Supplier TCI America Supplier No. C24701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55052-28-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD08272232
InChI Key HNTZVGMWXCFCTA-UHFFFAOYSA-N
Synonym 4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine
PubChem CID 11389493
IUPAC Name 4-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES ClC1=C2C=CNC2=NC=C1

Specifications

Melting Point 179°C
Quantity 1 g
Formula Weight 152.58
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-1H-pyrrolo[2,3-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.