Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,1,3-benzoxadiazole 98.0+%, TCI America™
SDP

Catalog No. C133725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C133725G 25 g
C13375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C133725G Supplier TCI America Supplier No. C133725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7116-16-7
Molecular Formula C6H3ClN2O
Molecular Weight (g/mol) 154.553
MDL Number MFCD00191329
InChI Key YLEBWUGKYMZPQQ-UHFFFAOYSA-N
Synonym 4-Chlorobenzofurazan
PubChem CID 592984
IUPAC Name 4-chloro-2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C(=C1)Cl

Specifications

Melting Point 82°C
Color Yellow
Quantity 25 g
Formula Weight 154.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,1,3-benzoxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.