Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,6-pyridinedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C29245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29245G Supplier TCI America Supplier No. C29245G

Chemical Identifiers

CAS 4722-94-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD04117894
InChI Key IYUMNONNHYADBU-UHFFFAOYSA-N
PubChem CID 304010
IUPAC Name 4-chloropyridine-2,6-dicarboxylic acid
SMILES C1=C(C=C(N=C1C(=O)O)C(=O)O)Cl

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,6-pyridinedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.