Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorobenzonitrile, 98+%

Catalog No. AAL2016006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016006 5 g
AAL2016003 1 g
AAL2016014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2016006 Supplier Thermo Scientific Chemicals Supplier No. L2016006
Add to Cart

CAS: 57381-51-8 | C7H3ClFN | 155.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57381-51-8
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00143284
InChI Key JRDMGVGCATYZPW-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzonitrile, 4-chloro-2-fluoro-benzonitrile, benzonitrile, 4-chloro-2-fluoro, 4-chloro-2-fluorobenzenecarbonitrile, ncr dg bf, pubchem3424, 1-chloro-3-fluorobezene, acmc-1b0zj, ksc493o3h
PubChem CID 93655
IUPAC Name 4-chloro-2-fluorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)F)C#N

Specifications

Melting Point 58°C to 61°C
Quantity 5 g
UN Number UN3439
Beilstein 4231142
Formula Weight 155.56
Percent Purity ≥98%
Chemical Name or Material 4-Chloro-2-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 260-712-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.