Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorobenzylamine hydrochloride, 94%, Thermo Scientific™

Catalog No. AC290640100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC290640100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290640100 Supplier Thermo Scientific Chemicals Supplier No. 290640100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 202982-63-6
CAS Min % 92.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 94%
Molecular Formula C7H8Cl2FN
Quantity 10g
MDL Number MFCD00143286
Synonym 4-chloro-2-fluorobenzylamine hydrochloride, 4-chloro-2-fluorophenyl methanamine hydrochloride, 4-chloro-2-fluorobenzylamine hcl, 4-chloro-2-fluorophenyl methylamine, chloride, 1-4-chloro-2-fluorophenyl methanamine hydrochloride, pubchem3428, acmc-1cp68, 4-chloro-2-fluoro-benzylamine hydrochloride
InChI Key DFRJZBWKVAXYRV-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)F)CN.Cl
IUPAC Name (4-chloro-2-fluorophenyl)methanamine;hydrochloride
Molecular Weight (g/mol) 196.046
PubChem CID 2724909
Formula Weight 196.05
Percent Purity ≥92%
Chemical Name or Material 4-Chloro-2-fluorobenzylamine hydrochloride, 94%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.