Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-2'-hydroxyacetophenone 96.0+%, TCI America™
SDP

Catalog No. C296825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C296825G 25 g
C29685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C296825G Supplier TCI America Supplier No. C296825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6921-66-0
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00238557
InChI Key QCVSDCHNBNFJDQ-UHFFFAOYSA-N
PubChem CID 1051513
IUPAC Name 1-(4-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
Synonym 1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
Formula Weight 170.59
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Chloro-2′-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.