Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C32481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32481G 1 g
C3248200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32481G Supplier TCI America Supplier No. C32481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 762287-57-0
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.398
MDL Number MFCD02684317
InChI Key NZRRMTBNTSBIFH-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxyphenyl boronic acid, 4-chloro-2-methoxybenzeneboronic acid, 4-chloro-2-methoxy-phenyl boronic acid, boronic acid, 4-chloro-2-methoxyphenyl, acmc-209p2g, ksc495a8j, ablock ab-12-6332, 2-methoxy-4-chlorophenylboronic acid, 4-chloranyl-2-methoxy-phenyl boronic acid, 4-chloro-2-methyloxy phenyl boronic acid
PubChem CID 3858606
IUPAC Name (4-chloro-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)OC)(O)O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.40
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.