Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. AAL1077806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1077806 5 g
AAL1077814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1077806 Supplier Thermo Scientific Chemicals Supplier No. L1077806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23165-53-9
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.65
MDL Number MFCD00041089
InChI Key XTYLRVPBHHRTMS-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl isothiocyanate, 3-chloro-6-isothiocyanatotoluene, benzene, 4-chloro-1-isothiocyanato-2-methyl, 4-chloro-2-methylphenylisothiocyanate, 4-chloro-2-methyl-phenyl isothiocyanate, 2-methyl-4-chlorophenyl isothiocyanate, 4-chloro-2-methylbenzenisothiocyanate, acmc-20amlo, chlorotrimethylsilane tmcs, 4-chloro-2-methyl isothiocyanate
PubChem CID 90021
IUPAC Name 4-chloro-1-isothiocyanato-2-methylbenzene
SMILES CC1=CC(Cl)=CC=C1N=C=S

Specifications

Melting Point 33°C to 35°C
Quantity 5 g
Odor Irritating
UN Number UN2923
Beilstein 2963683
Boiling Point 88°C to 90°C (0.4 mmHg)
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylphenyl isothiocyanate
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 245-472-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.