Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-pyridinemethanol 98.0+%, TCI America™
SDP

Catalog No. C29505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29505G 5 g
C29501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29505G Supplier TCI America Supplier No. C29505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63071-10-3
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD07437885
InChI Key UEAIOHHGRGSGGJ-UHFFFAOYSA-N
Synonym 4-Chloro-2-(hydroxymethyl)pyridine
PubChem CID 2763167
IUPAC Name (4-chloropyridin-2-yl)methanol
SMILES C1=CN=C(C=C1Cl)CO

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 66°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.