Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-3'-fluoroacetophenone, 97%

Catalog No. AAH2637306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2637306 5 g
AAH2637303 1 g
AAH2637314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2637306 Supplier Thermo Scientific Chemicals Supplier No. H2637306
Add to Cart

CAS: 151945-84-5 | C8H6ClFO | 172.58 g/mol

4'-Chloro-3'-fluoroacetophenone is a halogenated benzene derivative used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloro-3′-fluoroacetophenone is a halogenated benzene derivative used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151945-84-5
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD04115858
InChI Key ATZHNDSADJDUPJ-UHFFFAOYSA-N
Synonym 1-4-chloro-3-fluorophenyl ethanone, 4'-chloro-3'-fluoroacetophenone, 4-chloro-3-fluoroacetophenone, 3-fluoro-4-chloroacetophenone, 1-4-chloro-3-fluoro-phenyl ethanone, 1-4-chloro-3-fluorophenyl ethan-1-one, ethanone, 1-4-chloro-3-fluorophenyl, intermediates-zcf02162, 1-acetyl-4-chloro-3-fluorobenzene, 4-acetyl-1-chloro-2-fluorobenzene
PubChem CID 3822077
IUPAC Name 1-(4-chloro-3-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C(F)=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 4'-Chloro-3'-fluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.