Learn More
4-Chloro-3-iodobenzoic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | 214°C to 216°C |
| Quantity | 1 g |
| Synonym | 3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50 |
| Formula Weight | 282.46 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Chloro-3-iodobenzoic acid |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.