Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. C116525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C116525G 25 g
C11655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C116525G Supplier TCI America Supplier No. C116525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16588-34-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007078
InChI Key HETBKLHJEWXWBM-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-3-nitro, chembl51664, 4-chloro-3-nitro-benzaldehyde, pubchem14274, acmc-1c7bn, 3nitro-4-chlorobenzaldehyde, asischem r36965, dsstox_cid_24846, dsstox_rid_80526
PubChem CID 85505
IUPAC Name 4-chloro-3-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC(C=O)=CC=C1Cl

Specifications

Melting Point 62°C
Quantity 25 g
Formula Weight 185.56
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.