Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. C154825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154825G 25 g
C15485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C154825G Supplier TCI America Supplier No. C154825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 939-80-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00016987
InChI Key XBLPHYSLHRGMNW-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
PubChem CID 13655
IUPAC Name 4-chloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl

Specifications

Melting Point 99°C
Quantity 25 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzonitrile
EINECSNumber (3)-1781
RTECSNumber DI3050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.