Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzophenone 98.0+%, TCI America™
SDP

Catalog No. C290625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C290625G 25 g
C29065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C290625G Supplier TCI America Supplier No. C290625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56107-02-9
Molecular Formula C13H8ClNO3
Molecular Weight (g/mol) 261.66
MDL Number MFCD00007081
InChI Key YBDBYPQFIMSFJW-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
PubChem CID 41743
IUPAC Name (4-chloro-3-nitrophenyl)(phenyl)methanone
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1

Specifications

Melting Point 106°C
Color Yellow
Boiling Point 235°C
Quantity 25 g
Formula Weight 261.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzophenone
RTECSNumber DJ0425000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.