Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. C022525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022525G 25 g
C0225500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C022525G Supplier TCI America Supplier No. C022525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121-17-5
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
PubChem CID 8462
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 2307
Formula Weight 225.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride
EINECSNumber (3)-0452
RTECSNumber XS9140000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.