Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C32121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32121G 1 g
C3212-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32121G Supplier TCI America Supplier No. C32121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151169-67-4
Molecular Formula C6H5BClNO4
Molecular Weight (g/mol) 201.37
MDL Number MFCD02258950
InChI Key LQCCRPUZTXKDGB-UHFFFAOYSA-N
PubChem CID 5006651
IUPAC Name (4-chloro-3-nitrophenyl)boronic acid
SMILES OB(O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Specifications

Melting Point 275°C
Quantity 1 g
Synonym 4-chloro-3-nitrophenyl boronic acid, 4-chloro-3-nitrobenzeneboronic acid, 3-nitro-4-chlorophenylboronic acid, 5-borono-2-chloronitrobenzene, 4-chloro-3-nitro-phenyl boronic acid, boronic acid,b-4-chloro-3-nitrophenyl, boronic acid, 4-chloro-3-nitrophenyl, 4-chloro-3-nitrophenylboronicacid, pubchem1784, akos brn-0709
Formula Weight 201.37
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.