Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-4'-hydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. C115525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C115525G 25 g
C1155500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C115525G Supplier TCI America Supplier No. C115525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42019-78-3
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.66
MDL Number MFCD00002357
InChI Key RUETVLNXAGWCDS-UHFFFAOYSA-N
Synonym 4-chloro-4'-hydroxybenzophenone, 4-chlorophenyl 4-hydroxyphenyl methanone, 4-hydroxy-4'-chlorobenzophenone, 4-p-chlorobenzoylphenol, 4-4-chlorobenzoyl phenol, 4-chlorophenyl 4-hydroxyphenyl-methanone, methanone, 4-chlorophenyl 4-hydroxyphenyl, 4-chlorophenyl-4-hydroxyphenyl methanone, 4-chlorophenyl 4-hydroxyphenyl ketone, pubchem3317
PubChem CID 123504
IUPAC Name 4-(4-chlorobenzoyl)phenol
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 180°C
Quantity 25 g
Formula Weight 232.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-4′-hydroxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.