Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic Acid 99.0+%, TCI America™
SDP

Catalog No. C13235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13235G 5 g
C13231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13235G Supplier TCI America Supplier No. C13235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 50892-23-4 | C14H14ClN3O2S | 323.80 g/mol

Chemical Identifiers

CAS 50892-23-4
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00191335
InChI Key SZRPDCCEHVWOJX-UHFFFAOYSA-N
Synonym pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio
PubChem CID 5694
ChEBI CHEBI:32509
IUPAC Name 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid
SMILES CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 323.80
Percent Purity ≥99.0% (HPLC,T)
Grade Not Available
Chemical Name or Material [4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic Acid
RTECSNumber AG2915000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.