Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-iodoquinazoline 98.0+%, TCI America™
SDP

Catalog No. C27751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27751G Supplier TCI America Supplier No. C27751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98556-31-1
Molecular Formula C8H4ClIN2
Molecular Weight (g/mol) 290.49
MDL Number MFCD01862193
InChI Key BDAIUOPDSRAOKI-UHFFFAOYSA-N
Synonym 4-chloro-6-iodo-quinazoline, quinazoline, 4-chloro-6-iodo, 6-iodo-4-chloroquinazoline, pubchem23075, 4chloro-6-iodoquinazoline, 6-iodo4-chloroquinazoline, 4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-qulinazoline, 6-iodine-4-chloroquine thiazole moiety
PubChem CID 11173809
IUPAC Name 4-chloro-6-iodoquinazoline
SMILES ClC1=NC=NC2=CC=C(I)C=C12

Specifications

Melting Point 177°C
Color Yellow
Quantity 1 g
Formula Weight 290.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6-iodoquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.