Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-methoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. C23691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23691G 1 g
C23695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23691G Supplier TCI America Supplier No. C23691G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26452-81-3
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD06200211
InChI Key KLJGSQVYUGQOAW-UHFFFAOYSA-N
PubChem CID 1051509
IUPAC Name 4-chloro-6-methoxypyrimidine
SMILES COC1=CC(=NC=N1)Cl

Specifications

Color White
Boiling Point 80°C
Quantity 1 g
Formula Weight 144.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-6-methoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.