Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-methyl-2-(methylthio)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. C29521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29521G Supplier TCI America Supplier No. C29521G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17119-73-2
Molecular Formula C6H7ClN2S
Molecular Weight (g/mol) 174.646
MDL Number MFCD00023233
InChI Key ALMBOXQFPLQVLF-UHFFFAOYSA-N
Synonym 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine
PubChem CID 241163
IUPAC Name 4-chloro-6-methyl-2-methylsulfanylpyrimidine
SMILES CC1=CC(=NC(=N1)SC)Cl

Specifications

Melting Point 40°C
Boiling Point 147°C
Quantity 1 g
Formula Weight 174.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-6-methyl-2-(methylthio)pyrimidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.