Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-7-azaindole, 98%

Catalog No. AAH5673403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5673403 1 g
AAH56734MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5673403 Supplier Thermo Scientific Chemicals Supplier No. H5673403
Add to Cart

CAS: 55052-28-3 | C7H5ClN2 | 152.58 g/mol

A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55052-28-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD08272232
InChI Key HNTZVGMWXCFCTA-UHFFFAOYSA-N
Synonym 4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine
PubChem CID 11389493
IUPAC Name 4-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES ClC1=C2C=CNC2=NC=C1

Specifications

Melting Point 176°C to 181°C
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 152.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-7-azaindole
Hazard Category H301-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.