Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-alpha-methylbenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. C019810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C019810G 10 g
C019825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C019810G Supplier TCI America Supplier No. C019810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3391-10-4
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00041038
InChI Key MVOSNPUNXINWAD-UHFFFAOYNA-N
Synonym 1-4-chlorophenyl ethanol, 4-chloro-alpha-methylbenzyl alcohol, 1-p-chlorophenyl ethanol, 4-chlorophenylmethylcarbinol, 1-4-chlorophenyl ethan-1-ol, benzenemethanol, 4-chloro-.alpha.-methyl, 1-4'-chlorophenyl-1-hydroxyethane, benzyl alcohol, p-chloro-.alpha.-methyl, .alpha.-p-chlorophenyl ethanol, p-chloro-.alpha.-methylbenzyl alcohol
PubChem CID 18825
IUPAC Name 1-(4-chlorophenyl)ethanol
SMILES CC(C1=CC=C(C=C1)Cl)O

Specifications

Color Yellow
Quantity 10 g
Physical Form Clear Liquid at 20°C
Boiling Point 136°C
Formula Weight 156.61
Percent Purity ≥97.0% (GC)
Chemical Name or Material 4-Chloro-alpha-methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.