Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-DL-phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™

Catalog No. AAH6326006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6326006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6326006 Supplier Thermo Scientific Chemicals Supplier No. H6326006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14173-40-1
Molecular Formula C10H13Cl2NO2
Molecular Weight (g/mol) 250.119
MDL Number MFCD00012530
InChI Key GCBCWTWQAFLKJG-UHFFFAOYSA-N
Synonym 4-chloro-dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, h-dl-phe 4-cl-ome.hcl, dl-4-chlorophenylalanine methyl ester hydrochloride, dl-p-chlorophenylalanine methyl ester hydrochloride, 4-chloro-dl-phe-ome hcl, methyl 2-amino-3-4-chlorophenyl propanoate, chloride, 4-chloro-dl-phe-ome hydrochloride, pcpa methyl ester hcl, 4-chloro-dl-phe-omecl
PubChem CID 2733277
IUPAC Name methyl 2-amino-3-(4-chlorophenyl)propanoate;hydrochloride
SMILES COC(=O)C(CC1=CC=C(C=C1)Cl)N.Cl

Specifications

Melting Point 186°C to 189°C (decomposition)
Quantity 5 g
Formula Weight 250.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-DL-phenylalanine methyl ester hydrochloride
EINECSNumber 238-024-0
RTECSNumber AY4452100
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.