accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COONH3Cl

CAS RN 14173-40-1

IUPAC Name: 3-(4-chlorophenyl)-1-methoxy-1-oxopropan-2-aminium chloride
Synonyms: 3-(4-chlorophenyl)-1-methoxy-1-oxopropan-2-aminium chloride
Molecular Weight (g/mol): 250.12
Molecular Formula: C10H13Cl2NO2
InChi Key: GCBCWTWQAFLKJG-UHFFFAOYNA-N
SMILES: [Cl-].COC(=O)C([NH3+])CC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results