Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-o-xylene 98.0+%, TCI America™
SDP

Catalog No. C0356100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0356100G 100 g
C035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0356100G Supplier TCI America Supplier No. C0356100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 615-60-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000596
InChI Key HNQLMBJUMVLFCF-UHFFFAOYSA-N
Synonym 4-chloro-o-xylene, 3,4-dimethylchlorobenzene, 1-chloro-3,4-dimethylbenzene, benzene, 4-chloro-1,2-dimethyl, o-xylene, 4-chloro, 4-chloro-ortho-xylene, 4-chloro-1,2-xylene, unii-qo24waw04q, qo24waw04q, pubchem3641
PubChem CID 32886
IUPAC Name 4-chloro-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Cl)C

Specifications

Color Yellow
Quantity 100 g
Physical Form Clear Liquid at 20°C
Boiling Point 192°C
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.