Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloroacetophenone 97.0+%, TCI America™
SDP

Catalog No. C0033100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0033100G 100 g
C003325G 25 g
C0033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0033100G Supplier TCI America Supplier No. C0033100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 100 g
UN Number 3416
Formula Weight 154.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Chloroacetophenone
EINECSNumber (3)-1241
RTECSNumber KM5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.