Learn More
4-Chloroaniline, 98%
Description
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
Solubility
Soluble in water (2.6 g/Liter at 20°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
Specifications
Specifications
| Melting Point | 67°C to 72°C |
| Density | 1.43 |
| Quantity | 1000 g |
| UN Number | UN2018 |
| Beilstein | 471359 |
| Merck Index | 14,2118 |
| Boiling Point | 232°C |
| Solubility Information | Soluble in water (2.6g/Liter at 20°C ). |
| Formula Weight | 127.57 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H317-H350 |
| Hazard Statement | GHS H Statement H301-H311-H331-H350-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID |
| EINECSNumber | 203-401-0 |
| RTECSNumber | BX0700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.