Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzaldehyde, 98%

Catalog No. AC150441000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150441000 100 g Glass bottle
AC150442500 250 g Glass bottle
AC150440010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150441000 Supplier Thermo Scientific Chemicals Supplier No. 150441000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 104-88-1 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-88-1
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
InChI Key AVPYQKSLYISFPO-UHFFFAOYSA-N
Synonym p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
PubChem CID 7726
ChEBI CHEBI:28105
IUPAC Name 4-chlorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Cl

Specifications

Melting Point >46.0°C
Color White to Yellow
Density 1.1960g/mL
Boiling Point 214.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CHO
Quantity 100 g
Beilstein 07,235
Specific Gravity 1.196
Solubility Information Solubility in water: 935mg/l (20°C)
Formula Weight 140.57
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 4-Chlorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN:
EINECSNumber 203-247-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.