Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzamide 98.0+%, TCI America™
SDP

Catalog No. C0563250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0563250G 250 g
C056325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0563250G Supplier TCI America Supplier No. C0563250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 619-56-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007993
InChI Key BLNVISNJTIRAHF-UHFFFAOYSA-N
Synonym p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
PubChem CID 12084
IUPAC Name 4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 179°C
Color White
Quantity 250 g
Formula Weight 155.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzamide
EINECSNumber (3)-1430
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.