Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzoic acid, 99%

Catalog No. AC159250010
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159250010 1 kg Plastic bottle
AC159250250 25 g Plastic bottle
AC159252500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159250010 Supplier Thermo Scientific Chemicals Supplier No. 159250010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74-11-3 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-11-3
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00002531
InChI Key XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Synonym p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
PubChem CID 6318
ChEBI CHEBI:30747
IUPAC Name 4-chlorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color White to Yellow
Boiling Point 274.0°C to 276.0°C
Flash Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H4CO2H
Quantity 1 kg
Beilstein 09, 340
Merck Index 15, 2126
Solubility Information Solubility in water: 0,08g/L (20°C). Other solubilities: soluble in hot water, freely soluble in alcohol and ether
Formula Weight 156.57
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-805-9
RTECSNumber DG4976010
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.