Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C0141100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0141100G 100 g
C014125G 25 g
C0141500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0141100G Supplier TCI America Supplier No. C0141100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-01-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00000686
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
PubChem CID 8501
ChEBI CHEBI:60716
IUPAC Name 4-chlorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)Cl

Specifications

Color Colorless
Boiling Point 222°C
Quantity 100 g
UN Number 3265
Formula Weight 175.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
RTECSNumber DM6635510
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.