Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyryl chloride, 98%

Catalog No. AC108900010
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108900010 1 L
AC108902500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108900010 Supplier Thermo Scientific Chemicals Supplier No. 108900010
Add to Cart

CAS: 4635-59-0 | C4H6Cl2O | 141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4635-59-0
Molecular Formula C4H6Cl2O
Molecular Weight (g/mol) 141
MDL Number MFCD00000754
InChI Key CDIIZULDSLKBKV-UHFFFAOYSA-N
Synonym 4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro
PubChem CID 78370
IUPAC Name 4-chlorobutanoyl chloride
SMILES C(CC(=O)Cl)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -47.0°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 173.0°C to 174.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4605 to 1.4625
Linear Formula Cl(CH2)3COCl
Quantity 1 L
Beilstein 02, 278
Specific Gravity 1.25
Solubility Information Solubility in water: reacts.
Formula Weight 141
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyryl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear eye protection/face protecti
EINECSNumber 225-059-1
RTECSNumber EM1406000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.