Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloromandelic Acid 98%, Thermo Scientific™

Catalog No. AC136480250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC136480250 25 g
AC136481000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC136480250 Supplier Thermo Scientific Chemicals Supplier No. 136480250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-86-4
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
InChI Key BWSFWXSSALIZAU-UHFFFAOYSA-N
Synonym 4-chloromandelic acid, 2-4-chlorophenyl-2-hydroxyacetic acid, p-chloromandelic acid, p-chloro mandelic acid, 4-chloro-dl-mandelic acid, 4-chloromandelic acid, pract., benzeneacetic acid, 4-chloro-.alpha.-hydroxy, dl-4-chloromandelic acid, mandelic acid, p-chloro, 4-chlorophenyl hydroxy acetic acid
PubChem CID 10299
IUPAC Name 2-(4-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)Cl

Specifications

Melting Point 116°C to 120°C
Color White
CAS Min % 97.5
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Physical Form Crystalline Powder
Linear Formula ClC6H4CH(OH)COOH
Formula Weight 186.59
Percent Purity ≥97.5%
Chemical Name or Material 4-Chloromandelic acid, 98%
Show More Show Less
EINECSNumber 207-764-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.