Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloromethyl-5-methyl-1,3-dioxol-2-one 94.0+%, TCI America™
SDP

Catalog No. C188225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C188225G 25 g
C18825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C188225G Supplier TCI America Supplier No. C188225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80841-78-7
Molecular Formula C5H5ClO3
Molecular Weight (g/mol) 148.542
MDL Number MFCD07787494
InChI Key QCLFSYYUWPUWQR-UHFFFAOYSA-N
Synonym 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, pubchem10542, acmc-209pk7, ksc490e4r, 4-chloromethyl-5methyl-1,3-dioxol-2-one
PubChem CID 9855518
IUPAC Name 4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
SMILES CC1=C(OC(=O)O1)CCl

Specifications

Color Yellow
Boiling Point 93°C
Quantity 25 g
Formula Weight 148.54
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloromethyl-5-methyl-1,3-dioxol-2-one
EINECSNumber (5)-6296
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.