Learn More
4-(Chloromethyl)phenyltrichlorosilane, 97%
Description
4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
Specifications
Specifications
| Boiling Point | 142°C to 144°C (15mmHg) |
| Quantity | 25 g |
| UN Number | UN2987 |
| Beilstein | 3090295 |
| Flash Point | 78°C (172°F) |
| Solubility Information | Reacts with water. |
| Formula Weight | 260.02 |
| Percent Purity | 97% |
| Refractive Index | 1.548 |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Show More |
Safety and Handling
| Hazard Category | H227-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| EINECSNumber | 237-203-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.