Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenol, 99%

Catalog No. aaa1560222
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1560222 100 g
AAA1560230 250 g
AAA156020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1560222 Supplier Thermo Scientific Chemicals Supplier No. A1560222
Only null left
Add to Cart

CAS: 106-48-9 | C6H5ClO | 128.555 g/mol

It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

Solubility
Soluble in water (27g/L).

Notes
Air sensitive. Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-48-9
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002318
InChI Key WXNZTHHGJRFXKQ-UHFFFAOYSA-N
Synonym p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
PubChem CID 4684
ChEBI CHEBI:28078
IUPAC Name 4-chlorophenol
SMILES C1=CC(=CC=C1O)Cl

Specifications

Melting Point 39°C to 43°C
Density 1.306
Boiling Point 217°C to 220°C
Flash Point 115°C (239°F)
Odor Phenol-like
Quantity 100 g
UN Number UN2020
Beilstein 507004
Sensitivity Air Sensitive
Merck Index 14,2154
Solubility Information Soluble in water (27g/L).
Formula Weight 128.56
Percent Purity 99%
Chemical Name or Material 4-Chlorophenol
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 203-402-6
RTECSNumber SK2800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.