Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl Chlorothionoformate 98.0+%, TCI America™
SDP

Catalog No. C158825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C158825G 25 g
C15885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C158825G Supplier TCI America Supplier No. C158825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 937-64-4
Molecular Formula C7H4Cl2OS
Molecular Weight (g/mol) 207.068
MDL Number MFCD00014465
InChI Key BQIABQCJXBELMT-UHFFFAOYSA-N
Synonym 4-Chlorophenyl Chlorothioformate, Chlorothioformic Acid O-(4-Chlorophenyl) Ester
PubChem CID 70306
IUPAC Name O-(4-chlorophenyl) chloromethanethioate
SMILES C1=CC(=CC=C1OC(=S)Cl)Cl

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 g
UN Number 3265
Formula Weight 207.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl Chlorothionoformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.