Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl isocyanate, 98%

Catalog No. AC166565000
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166565000 500 g Glass bottle
AC166560250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166565000 Supplier Thermo Scientific Chemicals Supplier No. 166565000

missing translation for 'validMainframeMsg'

CAS: 104-12-1 | C7H4ClNO | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-12-1
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00002024
InChI Key ADAKRBAJFHTIEW-UHFFFAOYSA-N
Synonym 4-chlorophenyl isocyanate, p-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato, pcpi, 4-chloroisocyanatobenzene, para-chlorophenyl isocyanate, p-chlorfenylisokyanat, 4-chlorophenylisocyanate, isocyanic acid, p-chlorophenyl ester, 1-chloro-4-isocyanato-benzene
PubChem CID 7693
IUPAC Name 1-chloro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Cl

Specifications

Melting Point 29.0°C to 31.0°C
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Density 1.2600g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Refractive Index 1.5618
Specific Gravity 1.26
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4NCO
Beilstein 12, 616
Boiling Point 203.0°C to 204.0°C
Solubility Information Solubility in water: decomposes in water
Formula Weight 153.57
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if in
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 203-176-9
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.