Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl Phenyl Sulfone 96.0+%, TCI America™
SDP

Catalog No. C145125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C145125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C145125G Supplier TCI America Supplier No. C145125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80-00-2
Molecular Formula C12H9ClO2S
Molecular Weight (g/mol) 252.712
MDL Number MFCD00007549
InChI Key OFCFYWOKHPOXKF-UHFFFAOYSA-N
Synonym 4-Chlorodiphenyl Sulfone, 1-Chloro-4-(phenylsulfonyl)benzene, Sulferone
PubChem CID 6621
IUPAC Name 1-(benzenesulfonyl)-4-chlorobenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 92°C
Color White
Boiling Point 389°C
Quantity 25 g
Formula Weight 252.71
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenyl Phenyl Sulfone
EINECSNumber (3)-0061
RTECSNumber WR5775000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.