Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl Phosphorodichloridate 97.0+%, TCI America™
SDP

Catalog No. C097725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097725G 25 g
C09775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C097725G Supplier TCI America Supplier No. C097725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 772-79-2
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009705
InChI Key CCZMQYGSXWZFKI-UHFFFAOYSA-N
Synonym 4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
PubChem CID 69879
IUPAC Name 1-chloro-4-dichlorophosphoryloxybenzene
SMILES C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 265°C
Quantity 25 g
UN Number 3265
Formula Weight 245.42
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl Phosphorodichloridate [Phosphorylating Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.