Learn More
4-Chlorophenylacetyl chloride, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Yellow |
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Density | 1.2920g/mL |
| Quantity | 25 g |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5495 to 1.5565 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Harmful if swallowed. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
| EINECSNumber | 246-571-1 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.