Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropyrazole 98.0+%, TCI America™
SDP

Catalog No. C288825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C288825G 25 g
C28885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C288825G Supplier TCI America Supplier No. C288825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15878-00-9
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.521
MDL Number MFCD00159634
InChI Key BADSZRMNXWLUKO-UHFFFAOYSA-N
Synonym 4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
PubChem CID 27524
IUPAC Name 4-chloro-1H-pyrazole
SMILES C1=C(C=NN1)Cl

Specifications

Melting Point 77°C
Color White
Boiling Point 220°C
Quantity 25 g
Formula Weight 102.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloropyrazole
RTECSNumber UQ6445000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.