Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorosalicylaldehyde 95.0+%, TCI America™
SDP

Catalog No. C26531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26531G 1 g
C26535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26531G Supplier TCI America Supplier No. C26531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2420-26-0
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD06252499
InChI Key QNZWAJZEJAOVPN-UHFFFAOYSA-N
Synonym 4-Chloro-2-hydroxybenzaldehyde
PubChem CID 520101
IUPAC Name 4-chloro-2-hydroxybenzaldehyde
SMILES OC1=C(C=O)C=CC(Cl)=C1

Specifications

Melting Point 47°C
Quantity 1 g
Formula Weight 156.57
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorosalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.